This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03957040_s1
See other MEG3 GE Assays ›
Gene Symbol:
Meg3
Gene Name:
maternally expressed 3
Gene Aliases:
2900016C05Rik, 3110050O07Rik, 6330408G06Rik, AI425946, AW108224, D12Bwg1266e, Gtl2, R74756, R75394
Chromosome Location:
Chr.12: 109540996 - 109571729 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03957040_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Meg3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_003633.3 5 11029 99
NR_027651.2 9 11017 99
GenBank mRNA AK019336.1 - 1 440 99
BP771368.1 - 1 99
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BE984512.1 - 1 Chr. 12 - 109568068 - 109568589 [-] on Build GRCm38 99
GenBankmRNA AI414995.1 - 1 Chr. 12 - 109567891 - 109568289 [-] on Build GRCm38 99
GenBankmRNA AW045449.1 - 1 Chr. 12 - 109567905 - 109568309 [-] on Build GRCm38 99
GenBankmRNA BF457080.1 - 1 Chr. 12 - 109568125 - 109568305 [-] on Build GRCm38 99
GenBankmRNA CX729634.1 - 1 557 Chr. 12 - 109567651 - 109568267 [+] on Build GRCm38 99
GenBankmRNA AW048823.1 - 1 Chr. 12 - 109567937 - 109568309 [-] on Build GRCm38 99
GenBankmRNA CB519316.1 - 1 574 Chr. 12 - 109567635 - 109568407 [+] on Build GRCm38 99
GenBankmRNA BQ442837.1 - 333 Chr. - - [-] on Build 99
GenBankmRNA BM114546.2 - 1 Chr. 12 - 109568069 - 109568648 [-] on Build GRCm38 99
GenBankmRNA AW049612.1 - 1 Chr. 12 - 109567997 - 109568309 [-] on Build GRCm38 99
GenBankmRNA BE983674.1 - 1 Chr. 12 - 109568113 - 109568589 [-] on Build GRCm38 99
GenBankmRNA BE945590.1 - 1 Chr. 12 - 109567946 - 109568309 [-] on Build GRCm38 99
GenBankmRNA CF743770.1 - 198 Chr. - - [-] on Build 99
GenBankmRNA CR520901.1 - 1 Chr. 12 - 109568113 - 109568589 [-] on Build GRCm38 99
GenBankmRNA CD775465.1 - 1 Chr. 12 - 109567786 - 109568309 [-] on Build GRCm38 99

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream microRNA 770 Mir770 Mirn770,
mir-770,
mmu-mir-770
-4.0kb Sense Chr. 12 - 109563692 - 109563785 on NCBI Build GRCm38
Downstream microRNA 673 Mir673 Mirn673,
mir-673,
mmu-mir-673
3.7kb Sense Chr. 12 - 109571990 - 109572080 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s)

in utero embryonic development
liver development
DNA methylation
skeletal muscle tissue development
determination of adult lifespan
post-embryonic development
multicellular organism growth
lung alveolus development
SMAD protein signal transduction
genetic imprinting

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200008099.mm.1 NR_003633.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200008099.mm.1 NR_027651.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200008099.mm.1 TC1200001127.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200008097.mm.1 NR_003633.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200008097.mm.1 NR_027651.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200008097.mm.1 TC1200001127.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200008096.mm.1 NR_003633.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200008096.mm.1 NR_027651.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200008096.mm.1 TC1200001127.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 671603 - -
ABI Mouse Genome Survey Microarray 802206 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline