This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03957759_s1
See other 5930409G06RIK GE Assays ›
Gene Symbol:
5930409G06Rik
Gene Name:
RIKEN cDNA 5930409G06 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03957759_s1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer: 407.65
407.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 5930409G06Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK020028.1 - 1 146 68
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY718015.1 - 1 146 Chr. 3 - 45274066 - 45274967 [-] on Build GRCm38 68

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream DNA segment,
Chr 3,
ERATO Doi 751,
expressed
D3Ertd751e 2810009O15Rik,
4930415G15Rik
-3.5mb Anti-sense Chr. 3 - 41742606 - 41803292 on NCBI Build GRCm38
Downstream RIKEN cDNA 2610316D01 gene 2610316D01Rik 5.5kb Sense Chr. 3 - 45280439 - 45378260 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2399529 AK020028.1 mouse_transcriptome
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1430745_PM_at - -
Mouse Genome 430 2.0 Array 1430745_at - -
Mouse Genome 430 2.0 Array 1430745_PM_at - -
Mouse Expression Set 430 1430745_PM_at - -
Mouse Expression Set 430 1430745_at - -
Affymetrix HT Mouse Genome 430B Array 1430745_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1430745_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 856990 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline