Assay Details
Gene Symbol: 2010107E04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK020400.1 | - | 1 | 242 | 100 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
predicted gene, 36757 |
Gm36757 | -8.8kb | Anti-sense | Chr. 12 - 111945958 - 111954147 on NCBI Build GRCm38 | |
Downstream | tudor domain containing 9 | Tdrd9 | 4930441E05Rik | 7.7kb | Anti-sense | Chr. 12 - 111971474 - 112068854 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
