This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03970145_m1
See other GM32122 GE Assays ›
Gene Symbol:
Gm32122
Gene Name:
predicted gene, 32122
Gene Aliases:
-
Chromosome Location:
Chr.8: 96371707 - 96404082 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03970145_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gm32122
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK134720.1 - 3 - 4 1661 91

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream glutamatic-oxaloacetic transaminase 2,
mitochondrial
Got2 AL022787,
FABP-pm,
Got-2,
mAspAT
-487.9kb Sense Chr. 8 - 95864137 - 95888365 on NCBI Build GRCm38
Downstream predicted gene 15679 Gm15679 OTTMUSG00000025218 2.6mb Anti-sense Chr. 8 - 99007127 - 99032740 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mus musculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-7669c5b596-vr462:80/100.66.78.194:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0