This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03975931_m1
See other GM29811 GE Assays ›
Gene Symbol:
Gm29811
Gene Name:
predicted gene, 29811
Gene Aliases:
-
Chromosome Location:
Chr.3: 132842202 - 132878596 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03975931_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gm29811
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_102297.1 1 - 2 589 71
GenBank mRNA AK135894.1 - 1 - 2 590 71
CF173244.1 - 1 - 2 579 71
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DV652548.1 - 1 - 2 569 Chr. 3 - 132843029 - 132878576 [-] on Build GRCm38 71
GenBankmRNA DV657469.1 - 1 - 2 577 Chr. 3 - 132843049 - 132878584 [-] on Build GRCm38 71
GenBankmRNA BQ560072.1 - 1 - 2 557 Chr. 3 - 132849293 - 132878564 [-] on Build GRCm38 71
GenBankmRNA DV651690.1 - 1 - 2 567 Chr. 3 - 132843353 - 132878574 [-] on Build GRCm38 71
GenBankmRNA DV658064.1 - 1 - 2 586 Chr. 3 - 132843360 - 132878581 [-] on Build GRCm38 71
GenBankmRNA BY719819.1 - 1 - 2 587 Chr. 3 - 132849246 - 132878594 [-] on Build GRCm38 71

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream TBC1 domain containing kinase Tbck 1700120J03Rik,
9430001M19,
A630047E20Rik,
C030007I09Rik,
Tbckl
-7.5kb Anti-sense Chr. 3 - 132683994 - 132841688 on NCBI Build GRCm38
Downstream nephronectin Npnt 1110009H02Rik,
AA682063,
AI314031,
Nctn,
POEM
3.1kb Sense Chr. 3 - 132881745 - 132950291 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300002971.mm.1

Set Membership:

Amplicon length between 71 and 85 Mus musculus Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300023432.mm.1 NR_102297.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300023432.mm.1 NR_102298.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300023432.mm.1 TC0300002971.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300023434.mm.1 NR_102297.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300023434.mm.1 NR_102298.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300023434.mm.1 TC0300002971.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline