This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03976736_m1
See other GM31077 GE Assays ›
Gene Symbol:
Gm31077
Gene Name:
predicted gene, 31077
Gene Aliases:
-
Chromosome Location:
Chr.16: 23515291 - 23560080 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03976736_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gm31077
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK144897.1 - 1 - 2 159 102
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY269008.1 - 1 - 2 187 Chr. 16 - 23520291 - 23610077 [+] on Build GRCm38 102
GenBankmRNA BY085263.1 - 1 - 2 269 Chr. 16 - 23520209 - 23527588 [+] on Build GRCm38 102
GenBankmRNA BY089916.1 - 1 - 2 266 Chr. 16 - 23520211 - 23527601 [+] on Build GRCm38 102
GenBankmRNA BY734578.1 - 1 - 2 159 Chr. 16 - 23520319 - 23543821 [+] on Build GRCm38 102

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream receptor transporter protein 1 Rtp1 Gm1766,
Gm604
-86.4kb Sense Chr. 16 - 23428601 - 23433960 on NCBI Build GRCm38
Downstream receptor transporter protein 4 Rtp4 5830458K16Rik,
Ifrg28
73.8kb Sense Chr. 16 - 23601379 - 23614222 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline