This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03979951_m1
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Species:
Mouse
Assay Type:
Non-coding
Assay ID Mm03979951_m1
Size
Availability Made To Order
Catalog # 4426961
Price 4,565.00
Your Price
Online offer:
4565.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BX523414.1 - 2 - 3 274 Chr. 3 - 98382461 - 98743923 [+] on Build GRCm38 105
GenBankmRNA BC002224.1 - 302 Chr. - - [-] on Build 105
GenBankmRNA BE308109.1 - 2 - 3 282 Chr. 3 - 98382459 - 98753401 [+] on Build GRCm38 105

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream hydroxy-delta-5-steroid dehydrogenase,
3 beta- and steroid delta-isomerase 5
Hsd3b5 -34.0kb Anti-sense Chr. 3 - 98618636 - 98645534 on NCBI Build GRCm38
Downstream hydroxy-delta-5-steroid dehydrogenase,
3 beta- and steroid delta-isomerase 3
Hsd3b3 9030618K22Rik,
AI790201
27.9kb Anti-sense Chr. 3 - 98741476 - 98763128 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0