This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03990338_m1
See other UCMA GE Assays ›
Gene Symbol:
Ucma
Gene Name:
upper zone of growth plate and cartilage matrix associated
Gene Aliases:
1110017I16Rik, AW121955, Grp
Chromosome Location:
Chr.2: 4976122 - 4985748 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03990338_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ucma
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001113558.2 NP_001107030.1 3 - 4 350 103
NM_026754.3 NP_081030.1 2 - 3 284 103
GenBank mRNA AK003750.1 - 2 - 3 226 103
BC115608.2 - 3 - 4 253 40102702 103
BC115609.2 - 3 - 4 253 40102703 103
EF529510.1 - 3 - 4 321 103
EU368184.1 - 3 - 4 350 103
FJ217399.1 - 3 - 4 288 103
FJ217400.1 - 2 - 3 222 103

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200000040.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ucma

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of osteoblast differentiation
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200000321.mm.1 NM_001113558.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200000321.mm.1 NM_001113558.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200000321.mm.1 NM_026754.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200000321.mm.1 NM_026754.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200000321.mm.1 TC0200000040.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hwq2n:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0