This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04179181_m1
See other FREM3 GE Assays ›
Gene Symbol:
Frem3
Gene Name:
Fras1 related extracellular matrix protein 3
Gene Aliases:
Gm795, NV2
Chromosome Location:
Chr.8: 80611039 - 80695557 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04179181_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Frem3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001167898.1 NP_001161370.1 6 - 7 6048 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800000890.mm.1

Set Membership:

Amplicon length less than or equal to 70 Transporter Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

cell communication
cell adhesion
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800006644.mm.1 NM_001167898.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800006644.mm.1 NM_001167898.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800006644.mm.1 TC0800000890.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hwq2n:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0