This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04180196_m1
See other TMEM91 GE Assays ›
Gene Symbol:
Tmem91
Gene Name:
transmembrane protein 91
Gene Aliases:
A830041P22Rik, DSPC3
Chromosome Location:
Chr.7: 25669139 - 25679815 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04180196_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmem91
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_177102.4 NP_796076.1 3 - 4 514 103
GenBank mRNA AK079312.1 - 3 - 4 515 103
BC058996.1 - 3 - 4 463 5711063 103

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700002641.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tmem91

Gene Ontology Categories:

Function(s) Process(es)

hematopoietic progenitor cell differentiation
response to biotic stimulus
molecular_function

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700022812.mm.1 TC0700002641.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700022812.mm.1 NM_177102.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700022812.mm.1 NM_177102.4 Refseq
Affymetrix Mouse Clariom S Array PSR0700022814.mm.1 TC0700002641.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700022814.mm.1 NM_177102.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700022814.mm.1 NM_177102.4 Refseq
Affymetrix Mouse Clariom S Array PSR0700022816.mm.1 TC0700002641.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700022816.mm.1 NM_177102.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700022816.mm.1 NM_177102.4 Refseq
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 435159 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0