This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04205839_m1
See other WRB GE Assays ›
Gene Symbol:
Wrb
Gene Name:
tryptophan rich basic protein
Gene Aliases:
5530402J05Rik, C030018G21Rik, Chd5
Chromosome Location:
Chr.16: 96145354 - 96189247 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04205839_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Wrb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_207301.2 NP_997184.1 3 - 4 385 111
GenBank mRNA AK021091.1 - 3 - 4 384 111
AK172079.1 - 3 - 4 398 111
BC031769.1 - 3 - 4 385 4235618 111

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600001079.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Wrb

Gene Ontology Categories:

Function(s) Process(es)

biological_process
tail-anchored membrane protein insertion into ER membrane
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600008536.mm.1 NM_207301.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600008536.mm.1 NM_207301.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600008536.mm.1 TC1600001079.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline