This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04206152_g1
See other GMNN GE Assays ›
Gene Symbol:
Gmnn
Gene Name:
geminin
Gene Aliases:
AW546347, Gem
Chromosome Location:
Chr.13: 24751845 - 24761937 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04206152_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gmnn
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_020567.2 NP_065592.1 2 - 3 182 68
GenBank mRNA AF068780.1 - 2 - 3 329 68
AK131959.1 - 2 - 3 156 68
BC020061.1 - 2 - 3 334 3988065 68

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gmnn

Gene Ontology Categories:

Function(s) Process(es)

regulation of DNA replication
protein complex assembly
cell cycle
negative regulation of DNA replication
organ morphogenesis
negative regulation of cell cycle
negative regulation of transcription, DNA-templated
transcription corepressor activity
protein binding
histone deacetylase binding
transcription regulatory region DNA binding
repressing transcription factor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300010366.mm.1 NM_020567.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300010366.mm.1 NM_020567.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300010366.mm.1 TC1300001666.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300010375.mm.1 NM_020567.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300010375.mm.1 NM_020567.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300010375.mm.1 TC1300001666.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300010376.mm.1 NM_020567.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300010376.mm.1 NM_020567.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300010376.mm.1 TC1300001666.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300010368.mm.1 NM_020567.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300010368.mm.1 NM_020567.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300010368.mm.1 TC1300001666.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300010372.mm.1 TC1300001666.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-s2cfr:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0