This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04207309_g1
See other ATP5G1 GE Assays ›
Gene Symbol:
Atp5g1
Gene Name:
ATP synthase, H+ transporting, mitochondrial F0 complex, subunit C1 (subunit 9)
Gene Aliases:
-
Chromosome Location:
Chr.11: 96072793 - 96075694 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04207309_g1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atp5g1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007506.6 NP_031532.2 1 - 2 62 97
GenBank mRNA BY297329.1 - 1 - 2 62 97

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_007506.6 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Transporter Detect Genomic DNA Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Atp5g1

Panther Classification:

Molecular Function -

ATP synthase

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
ATP synthesis coupled proton transport
ATP hydrolysis coupled proton transport
proton transport
ATP metabolic process
lipid binding
hydrogen ion transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100033087.mm.1 NM_007506.6.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100033087.mm.1 NM_007506.6 Refseq
Affymetrix Mouse Clariom S Array PSR1100033087.mm.1 TC1100003610.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0