This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04207428_g1
See other GAD1 GE Assays ›
Gene Symbol:
Gad1
Gene Name:
glutamate decarboxylase 1
Gene Aliases:
EP10, GAD25, GAD44, GAD67, Gad-1
Chromosome Location:
Chr.2: 70561763 - 70602014 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04207428_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gad1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008077.4 NP_032103.2 4 - 5 530 101
GenBank mRNA AF326547.1 - 4 - 5 480 101
AF326548.1 - 4 - 5 480 101
AF483492.1 - 3 - 4 297 101
AF483493.1 - 3 - 4 297 101
AK047521.1 - 4 - 5 666 101
AK054554.1 - 4 - 5 532 101
AK136801.1 - 4 - 5 515 101
AK148497.1 - 4 - 5 530 101
AK158827.1 - 3 - 4 296 101
AK160410.1 - 3 - 4 441 101
AK160868.1 - 4 - 5 528 101
BC027059.1 - 4 - 5 481 5358787 101
KC134210.1 - 3 - 4 297 101
KM102984.1 - 4 - 5 530 101
KM102985.1 - 4 - 5 665 101
Y12257.1 - 3 - 4 297 101
Z49976.1 - 4 - 5 417 101

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gad1

Panther Classification:

Molecular Function -

decarboxylase

Gene Ontology Categories:

Function(s) Process(es)

gamma-aminobutyric acid biosynthetic process
carboxylic acid metabolic process
social behavior
locomotory exploration behavior
neurotransmitter biosynthetic process
catalytic activity
glutamate decarboxylase activity
glutamate binding
lyase activity
carboxy-lyase activity
pyridoxal phosphate binding
protein heterodimerization activity
protein N-terminus binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200008190.mm.1 NM_008077.5 Refseq
Affymetrix Mouse Clariom S Array PSR0200008190.mm.1 TC0200001023.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0