This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04207652_gH
See other RAET1A GE Assays ›
Gene Symbol:
Raet1a Raet1d Raet1e
Gene Name:
retinoic acid early transcript 1, alpha
retinoic acid early transcript delta
retinoic acid early transcript 1E
Gene Aliases:
RAE-1alpha, Rae1alpha, Raet1
RAE-1delta
-
Chromosome Location:
-
Chromosome Location:
Chr.10: 22361865 - 22374143 on Build GRCm38
Chromosome Location:
Chr.10: 22158532 - 22191791 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
-
Assay ID Mm04207652_gH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Raet1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009016.1 NP_033042.1 3 - 4 282 94
GenBank mRNA D64160.1 - 3 - 4 282 94
Gene Symbol: Raet1d
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA FJ594066.1 - 222 94
Gene Symbol: Raet1e
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK131854.1 - 3 - 4 323 94
FJ594065.1 - 3 - 4 222 94
FJ594067.1 - 2 - 3 124 94

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the D64160.1,FJ594065.1,FJ594066.1,FJ594067.1,NM_009016.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Raet1a
Click here to find siRNAs for Gene Raet1d
Click here to find siRNAs for Gene Raet1e

Panther Classification:

Molecular Function -

major histocompatibility complex protein

Gene Ontology Categories:

Function(s) Process(es)

immune response
antigen processing and presentation
defense response to bacterium
cellular response to lipopolysaccharide
cellular response to exogenous dsRNA
natural killer cell lectin-like receptor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000001291.mm.1 NM_198193.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000001291.mm.1 NM_198193.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000001291.mm.1 TC1000000175.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline