This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04207768_g1
See other UBE2E3 GE Assays ›
Gene Symbol:
Ube2e3
Gene Name:
ubiquitin-conjugating enzyme E2E 3
Gene Aliases:
Ubce4, ubcM2
Chromosome Location:
Chr.2: 78868243 - 78920720 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04207768_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ube2e3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009454.2 NP_033480.1 1 - 2 363 92
GenBank mRNA AK048227.1 - 1 - 2 173 92
AK076011.1 - 1 - 2 170 92
AK168072.1 - 1 - 2 188 92
BC011477.1 - 1 - 2 363 4195671 92
X92664.1 - 1 - 2 147 92

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK076011.1,AK168072.1,BC011477.1,NM_009454.2,X92664.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ube2e3

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

protein ubiquitination
regulation of growth
protein K63-linked ubiquitination
protein K48-linked ubiquitination
protein K11-linked ubiquitination
nucleotide binding
ubiquitin-protein transferase activity
protein binding
ATP binding
ligase activity
ubiquitin conjugating enzyme activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200009238.mm.1 NM_009454.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200009238.mm.1 NM_009454.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200009238.mm.1 TC0200001150.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200009236.mm.1 NM_009454.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200009236.mm.1 NM_009454.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200009236.mm.1 TC0200001150.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200009240.mm.1 NM_009454.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200009240.mm.1 NM_009454.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200009240.mm.1 TC0200001150.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200009237.mm.1 NM_009454.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200009237.mm.1 NM_009454.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200009237.mm.1 TC0200001150.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 452130 - -
ABI Mouse Genome Survey Microarray 687253 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0