This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04207993_m1
See other GJB5 GE Assays ›
Gene Symbol:
Gjb5
Gene Name:
gap junction protein, beta 5
Gene Aliases:
Cnx31.1, Cx31.1, Gjb-5
Chromosome Location:
Chr.4: 127354809 - 127358164 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04207993_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gjb5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010291.3 NP_034421.1 1 - 2 142 86
GenBank mRNA AK082917.1 - 1 - 2 143 86
M91236.1 - 1 - 2 99 86

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK082917.1, M91236.1, NM_010291.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gjb5

Panther Classification:

Molecular Function -

gap junction

Gene Ontology Categories:

Function(s)

cell communication
trophoblast giant cell differentiation
spongiotrophoblast differentiation
labyrinthine layer morphogenesis

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2165364 NM_010291.3 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2165364 AK082917.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 579366 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 316640 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 731415 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P168046 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2165364 NM_010291.3 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2165364 AK082917.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P385639 NM_010291.3 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P385639 AK082917.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P385639 BC011148.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P385639 M91236.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284889 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284896 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284933 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284938 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284919 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284906 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284885 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284890 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284915 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284917 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284914 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284932 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284922 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284883 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284899 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284912 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284936 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284939 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284930 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284926 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284897 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284944 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284892 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284904 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284888 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284943 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284925 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284887 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284921 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284908 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284884 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284902 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284893 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284928 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284929 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284941 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284910 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P168046 - -
Whole Mouse Genome G4122A A_51_P385639 NM_010291.3 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P385639 AK082917.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P385639 BC011148.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P385639 M91236.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131061 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P168046 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03735746 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03452065 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03799635 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03799632 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284910 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284838 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284915 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284928 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284959 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284950 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284965 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284899 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284953 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284833 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284948 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284883 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284956 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284972 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284860 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284827 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284896 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284908 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284939 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284830 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284926 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284855 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284889 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284850 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284957 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284867 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284945 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284881 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284936 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284859 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284847 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284811 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284843 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284845 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284825 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284857 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284884 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284818 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284963 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284879 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284973 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284851 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284890 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284892 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284897 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284812 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284922 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284885 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284882 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284930 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284846 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284824 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284864 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284840 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284895 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284946 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284969 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284904 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284852 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284822 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284893 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284943 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284828 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284831 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284917 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284921 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284841 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284873 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284862 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284865 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284933 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284925 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284977 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284914 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284876 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284960 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284919 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284870 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284888 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284836 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284861 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284906 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284902 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284933 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284908 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284883 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284929 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284946 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284922 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03799639 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284941 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284934 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284904 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03689277 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284921 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284888 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284926 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284912 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284953 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284892 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284954 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284928 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03799633 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284965 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284914 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284925 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284969 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284973 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284889 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284948 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03634584 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03271233 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284945 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284895 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284893 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284917 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284957 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284897 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284938 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03452072 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284956 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284896 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03271239 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284944 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284915 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284899 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284939 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284886 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284890 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284935 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284970 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03557992 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284963 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284960 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284902 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284967 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284959 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284910 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23284972 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10516494 - -
Affymetrix Mouse Gene 1.0 ST Array 10516491 - -
Affymetrix Mouse Gene 1.0 ST Array 10516492 - -
Affymetrix Mouse Gene 1.0 ST Array 10516493 - -
Affymetrix HT MG-430 PM Array Plate 1449204_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10516490 - -
Mouse Genome 430 2.0 Array 1449204_PM_at - -
Mouse Genome 430 2.0 Array 1449204_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 14622_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 14622_at - -
Affymetrix Mouse Clariom S Array PSR0400029045.mm.1 NM_010291.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400029045.mm.1 NM_010291.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400029045.mm.1 TC0400003568.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400029043.mm.1 NM_010291.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400029043.mm.1 NM_010291.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400029043.mm.1 TC0400003568.mm.1 MTA TC
Mouse Expression Set 430 1449204_PM_at - -
Mouse Expression Set 430 1449204_at - -
Murine Genome U74v2 Set 104016_at - -
Murine Genome U74v2 Set 104016_PM_at - -
Mouse Genome 430A 2.0 Array 1449204_at - -
Mouse Genome 430A 2.0 Array 1449204_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1449204_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1449204_at - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 3440022 - -
Illumina mouse-6 v1.0 expression beadchip 3440022 - -
Illumina mouseRef-8 v1.0 expression beadchip 0003440022 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2766772 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2766772 - -
mouse-6 v1.1 (Illumina) scl23801.2_390-S - -
Illumina Sentrix mouse-6.v1 bead array 3440022 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl23801.2_390-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 3440022 - -
MouseRef-8 v2 Expression BeadChip ILMN_2766772 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2766772 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline