This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04208000_g1
See other GTF2H4 GE Assays ›
Gene Symbol:
Gtf2h4
Gene Name:
general transcription factor II H, polypeptide 4
Gene Aliases:
AW545633, BTF2 p52, p52
Chromosome Location:
Chr.17: 35667728 - 35673750 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04208000_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gtf2h4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AF054823.1 - 2 - 3 265 111
AK079456.1 - 2 - 3 294 111
AK088296.1 - 2 - 3 294 111
AK144680.1 - 2 - 3 296 111
AK146112.1 - 2 - 3 295 111
AK149320.1 - 2 - 3 296 111
BC012638.1 - 1 - 2 154 4191809 111

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

RNA metabolism protein nucleic acid metabolism protein general transcription factor

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700018781.mm.1 NM_010364.4 Refseq
Affymetrix Mouse Clariom S Array PSR1700018781.mm.1 TC1700001975.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700018779.mm.1 NM_010364.4 Refseq
Affymetrix Mouse Clariom S Array PSR1700018779.mm.1 TC1700001975.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0