This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04208139_g1
See other SEC22B GE Assays ›
Gene Symbol:
Sec22b
Gene Name:
SEC22 vesicle trafficking protein homolog B (S. cerevisiae)
Gene Aliases:
4930564D15Rik, AA517334, AI480645, C81333, ERS-24, Sec22l1
Chromosome Location:
Chr.3: 97901227 - 97922318 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04208139_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sec22b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011342.4 NP_035472.1 2 - 3 301 108
GenBank mRNA AK010363.1 - 2 - 3 310 108
AK017237.1 - 2 - 3 307 108
AK034973.1 - 2 - 3 313 108
AK037918.1 - 2 - 3 302 108
AK088514.1 - 2 - 3 301 108
AK089928.1 - 2 - 3 294 108
AK134215.1 - 2 - 3 302 108
AK190268.1 - 1 - 2 101 108
BC009024.1 - 2 - 3 263 3481731 108
U91538.1 - 2 - 3 258 108

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sec22b

Gene Ontology Categories:

Function(s) Process(es)

transport
ER to Golgi vesicle-mediated transport
protein transport
vesicle-mediated transport
regulation of organelle organization
positive regulation of protein catabolic process
vesicle fusion with Golgi apparatus
SNARE binding
SNAP receptor activity
syntaxin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300007853.mm.1 NM_011342.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300007853.mm.1 NM_011342.4 Refseq
Affymetrix Mouse Clariom S Array PSR0300007853.mm.1 TC0300000972.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rhr4g:80/100.66.78.163:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0