This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04208293_g1
See other EIF6 GE Assays ›
Gene Symbol:
Eif6
Gene Name:
eukaryotic translation initiation factor 6
Gene Aliases:
1110004P11, AA408895, CAB, Itgb4bp, eIF-6, imc-415, p27(BBP), p27BBP
Chromosome Location:
Chr.2: 155819837 - 155826925 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04208293_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eif6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010579.2 NP_034709.1 2 - 3 684 95
GenBank mRNA AF047046.1 - 2 - 3 423 95
AF108462.2 - 2 - 3 469 95
AK003447.1 - 2 - 3 440 95
AK007594.1 - 2 - 3 451 95
AK012218.1 - 2 - 3 450 95
AK088707.1 - 2 - 3 447 95
AK134364.1 - 2 - 3 451 95
AK150400.1 - 2 - 3 458 95
AK150720.1 - 2 - 3 457 95
AK150931.1 - 2 - 3 459 95
AK151089.1 - 2 - 3 459 95
AK151102.1 - 2 - 3 442 95
AK151559.1 - 2 - 3 438 95
AK151644.1 - 2 - 3 458 95
AK152093.1 - 2 - 3 443 95
AK152215.1 - 2 - 3 443 95
AK152416.1 - 2 - 3 465 95
AK152832.1 - 2 - 3 443 95
AK159157.1 - 2 - 3 443 95
AK165946.1 - 2 - 3 242 95
AK167422.1 - 2 - 3 438 95
AK168257.1 - 2 - 3 459 95
BC015274.1 - 2 - 3 684 4219358 95
BC024442.1 - 2 - 3 412 4973875 95

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eif6

Panther Classification:

Molecular Function -

translation factor translation initiation factor translational protein

Gene Ontology Categories:

Function(s) Process(es)

translation
translational initiation
ribosome biogenesis
mature ribosome assembly
translation initiation factor activity
ribosome binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200042258.mm.1 NM_010579.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200042258.mm.1 NM_010579.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200042258.mm.1 TC0200004973.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0