This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04208295_g1
See other EIF6 GE Assays ›
Gene Symbol:
Eif6
Gene Name:
eukaryotic translation initiation factor 6
Gene Aliases:
1110004P11, AA408895, CAB, Itgb4bp, eIF-6, imc-415, p27(BBP), p27BBP
Chromosome Location:
Chr.2: 155819837 - 155826925 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04208295_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eif6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010579.2 NP_034709.1 4 - 5 1040 102
GenBank mRNA AF047046.1 - 4 - 5 779 102
AF108462.2 - 4 - 5 825 102
AK003447.1 - 4 - 5 796 102
AK007594.1 - 4 - 5 807 102
AK012218.1 - 4 - 5 806 102
AK088707.1 - 4 - 5 803 102
AK134364.1 - 4 - 5 807 102
AK150400.1 - 4 - 5 814 102
AK150720.1 - 4 - 5 813 102
AK150931.1 - 4 - 5 815 102
AK151089.1 - 4 - 5 815 102
AK151102.1 - 4 - 5 798 102
AK151644.1 - 4 - 5 814 102
AK152093.1 - 4 - 5 799 102
AK152215.1 - 4 - 5 799 102
AK152416.1 - 4 - 5 821 102
AK152832.1 - 4 - 5 799 102
AK159157.1 - 4 - 5 799 102
AK165946.1 - 4 - 5 598 102
AK167422.1 - 4 - 5 794 102
AK168257.1 - 4 - 5 815 102
AK212176.1 - 2 - 3 225 102
BC015274.1 - 4 - 5 1040 4219358 102
BC024442.1 - 4 - 5 768 4973875 102

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eif6

Panther Classification:

Molecular Function -

translation factor translation initiation factor translational protein

Gene Ontology Categories:

Function(s) Process(es)

translation
translational initiation
ribosome biogenesis
mature ribosome assembly
translation initiation factor activity
ribosome binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200042255.mm.1 NM_010579.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200042255.mm.1 NM_010579.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200042255.mm.1 TC0200004973.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0