Assay Details
| Gene Symbol: Spryd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025697.3 | NP_079973.1 | 3 - 4 | 649 | 103 | |
| GenBank mRNA | AK018044.1 | - | 3 - 4 | 527 | 103 | |
| AK018153.1 | - | 3 - 4 | 650 | 103 | ||
| AK018290.1 | - | 3 - 4 | 499 | 103 | ||
| AK137685.1 | - | 498 | 103 | |||
| AK167306.1 | - | 3 - 4 | 491 | 103 | ||
| AK169059.1 | - | 3 - 4 | 498 | 103 | ||
| BC037015.1 | - | 3 - 4 | 483 | 5344409 | 103 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Spryd7
Genomic Map