This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04208728_g1
See other NAA20 GE Assays ›
Gene Symbol:
Naa20
Gene Name:
N(alpha)-acetyltransferase 20, NatB catalytic subunit
Gene Aliases:
1500004D14Rik, 2900026I01Rik, AU041458, D2Ertd186e, Nat5
Chromosome Location:
Chr.2: 145903206 - 145916425 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04208728_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Naa20
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026425.3 NP_080701.1 3 - 4 274 115
GenBank mRNA AK005143.1 - 3 - 4 247 115
AK153145.1 - 3 - 4 256 115

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Naa20

Panther Classification:

Molecular Function -

acetyltransferase transferase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

N-terminal peptidyl-methionine acetylation
peptide alpha-N-acetyltransferase activity
N-acetyltransferase activity
transferase activity
transferase activity, transferring acyl groups

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200017154.mm.1 NM_001141965.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200017154.mm.1 NM_001141965.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200017154.mm.1 NM_026425.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200017154.mm.1 NM_026425.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200017154.mm.1 TC0200002170.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200017156.mm.1 NM_001141965.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200017156.mm.1 NM_001141965.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200017156.mm.1 NM_026425.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200017156.mm.1 NM_026425.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200017156.mm.1 TC0200002170.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200017155.mm.1 NM_001141965.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200017155.mm.1 NM_001141965.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200017155.mm.1 NM_026425.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200017155.mm.1 NM_026425.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200017155.mm.1 TC0200002170.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200017151.mm.1 NM_001141965.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200017151.mm.1 NM_001141965.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200017151.mm.1 NM_026425.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200017151.mm.1 NM_026425.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200017151.mm.1 TC0200002170.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200017152.mm.1 NM_001141965.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200017152.mm.1 NM_001141965.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200017152.mm.1 NM_026425.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200017152.mm.1 NM_026425.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200017152.mm.1 TC0200002170.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0