This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04210150_g1
See other RBM7 GE Assays ›
Gene Symbol:
Rbm7
Gene Name:
RNA binding motif protein 7
Gene Aliases:
1200007M24Rik, 1500011D06Rik, AU041934, AW554393
Chromosome Location:
Chr.9: 48488697 - 48495341 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04210150_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rbm7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_144948.5 NP_659197.2 3 - 4 427 95
NR_037589.1 4 - 5 532 95
GenBank mRNA AF458961.1 - 3 - 4 391 95
AK005200.1 - 3 - 4 403 95
AK006755.1 - 3 - 4 392 95
AK008645.1 - 3 - 4 387 95
AK011223.1 - 3 - 4 409 95
AK015662.1 - 3 - 4 428 95
AK145381.1 - 3 - 4 391 95
AK151056.1 - 3 - 4 406 95
AK151827.1 - 3 - 4 422 95
AK153504.1 - 4 - 5 496 95
BC054774.1 - 3 - 4 394 6807637 95
BC116931.1 - 3 - 4 376 40126250 95
BC119038.1 - 3 - 4 376 8733787 95

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rbm7

Panther Classification:

Molecular Function -

nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

biological_process
meiotic cell cycle
nucleotide binding
nucleic acid binding
RNA binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900018656.mm.1 NM_144948.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900018656.mm.1 NM_144948.5 Refseq
Affymetrix Mouse Clariom S Array PSR0900018656.mm.1 NR_037589.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900018656.mm.1 NR_037589.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900018656.mm.1 TC0900002236.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kt9qp:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0