This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04210283_m1
See other CMTR2 GE Assays ›
Gene Symbol:
Cmtr2
Gene Name:
cap methyltransferase 2
Gene Aliases:
AU022703, C730036L12Rik, Ftsjd1, MTr2
Chromosome Location:
Chr.8: 110215347 - 110224489 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04210283_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cmtr2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_146215.4 NP_666327.2 1 - 2 126 113
GenBank mRNA AK032855.1 - 1 - 2 102 113
AK050315.1 - 1 - 2 98 113
AK158368.1 - 1 - 2 108 113
BC025546.1 - 1 - 2 108 5320939 113

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK050315.1, AK158368.1, BC025546.1, NM_146215.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cmtr2

Gene Ontology Categories:

Function(s) Process(es)

7-methylguanosine mRNA capping
mRNA processing
methylation
cap2 mRNA methylation
mRNA (nucleoside-2'-O-)-methyltransferase activity
methyltransferase activity
transferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800010646.mm.1 NM_146215.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800010646.mm.1 NM_146215.4 Refseq
Affymetrix Mouse Clariom S Array PSR0800010646.mm.1 TC0800001316.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800010643.mm.1 NM_146215.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800010643.mm.1 NM_146215.4 Refseq
Affymetrix Mouse Clariom S Array PSR0800010643.mm.1 TC0800001316.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800010645.mm.1 NM_146215.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800010645.mm.1 NM_146215.4 Refseq
Affymetrix Mouse Clariom S Array PSR0800010645.mm.1 TC0800001316.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800010644.mm.1 NM_146215.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800010644.mm.1 NM_146215.4 Refseq
Affymetrix Mouse Clariom S Array PSR0800010644.mm.1 TC0800001316.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0