This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04211913_m1
See other PCED1B GE Assays ›
Gene Symbol:
Pced1b
Gene Name:
PC-esterase domain containing 1B
Gene Aliases:
Fam113b
Chromosome Location:
Chr.15: 97247080 - 97385696 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04211913_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pced1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172293.4 NP_758497.1 4 - 5 344 93
GenBank mRNA AK051480.1 - 4 - 5 345 93

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK051480.1, NM_172293.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pced1b

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500007547.mm.1 NM_172293.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500007547.mm.1 NM_172293.4 Refseq
Affymetrix Mouse Clariom S Array PSR1500007547.mm.1 TC1500000960.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0