Assay Details
| Gene Symbol: Snx33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175483.5 | NP_780692.2 | 1 - 2 | 3060 | 100 | |
| GenBank mRNA | AK048566.1 | - | 1 - 2 | 1565 | 100 | |
| AK053761.1 | - | 1 - 2 | 1595 | 100 | ||
| AK142274.1 | - | 1 - 2 | 3061 | 100 | ||
| BC096472.1 | - | 1 - 2 | 1810 | 3501525 | 100 | |
More Information
Set Membership: |
|

Snx33
Genomic Map