This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04213783_s1
See other PPP1R14C GE Assays ›
Gene Symbol:
Ppp1r14c
Gene Name:
protein phosphatase 1, regulatory (inhibitor) subunit 14c
Gene Aliases:
6330514J04Rik, KEPI
Chromosome Location:
Chr.10: 3366071 - 3464979 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04213783_s1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp1r14c
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133485.2 NP_597844.1 1 47 66
GenBank mRNA AF407167.2 - 1 143 66
AK047607.1 - 1 56 66
AK085997.1 - 1 85 66

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF407167.2, AK085997.1, NM_133485.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Inventoried Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppp1r14c

Panther Classification:

Molecular Function -

phosphatase modulator

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
regulation of phosphorylation
negative regulation of catalytic activity
protein phosphatase inhibitor activity
protein serine/threonine phosphatase inhibitor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000000008.mm.1 NM_133485.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000000008.mm.1 NM_133485.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000000008.mm.1 TC1000000002.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000000009.mm.1 NM_133485.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000000009.mm.1 NM_133485.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000000009.mm.1 TC1000000002.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-5h84w:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0