This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04213924_s1
See other CNTF GE Assays ›
Gene Symbol:
Cntf
Gene Name:
ciliary neurotrophic factor
Gene Aliases:
AI429687
Chromosome Location:
Chr.19: 12763528 - 12765632 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04213924_s1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cntf
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_170786.2 NP_740756.1 1 153 85
GenBank mRNA BC027539.1 - 1 152 1429801 85
W08735.1 - 1 105 85

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the BC027539.1,NM_170786.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cntf

Gene Ontology Categories:

Function(s) Process(es)

JAK-STAT cascade
multicellular organism development
nervous system development
positive regulation of cell proliferation
positive regulation of phosphatidylinositol 3-kinase signaling
cell differentiation
positive regulation of cell growth
growth
positive regulation of tyrosine phosphorylation of Stat3 protein
positive regulation of MAPK cascade
negative regulation of neuron apoptotic process
positive regulation of JAK-STAT cascade
negative regulation of photoreceptor cell differentiation
regulation of retinal cell programmed cell death
positive regulation of hormone secretion
astrocyte activation
muscle organ morphogenesis
neuron development
positive regulation of axon regeneration
protein heterooligomerization
regulation of resting membrane potential
membrane hyperpolarization
ciliary neurotrophic factor-mediated signaling pathway
negative regulation of neuron death
cytokine activity
ciliary neurotrophic factor receptor binding
interleukin-6 receptor binding
growth factor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0