This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04238115_s1
See other MIR132 GE Assays ›
Gene Symbol:
Mir132
Gene Name:
microRNA 132
Gene Aliases:
Mirn132, mir-132, mmu-mir-132
Chromosome Location:
Chr.11: 75173682 - 75173747 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04238115_s1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer: 407.65
407.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir132
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_029546.1 1 28 59

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream microRNA 212 Mir212 Mirn212,
mir-212,
mmu-mir-212
-204.0 Sense Chr. 11 - 75173388 - 75173478 on NCBI Build GRCm38
Downstream candidate tumor suppressor in ovarian cancer 2 Ovca2 9030414M07Rik 2.2kb Anti-sense Chr. 11 - 75175943 - 75178808 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s)

regulation of neuron maturation
regulation of dendrite development
response to oxygen levels
cellular response to inorganic substance

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.1 ST Array 10378547 - -
Affymetrix Mouse Clariom S Array PSR1100009577.mm.1 NR_029546.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100009577.mm.1 NR_029546.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100009577.mm.1 TC1100001062.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline