Assay Details
| Gene Symbol: 4930519F09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033601.1 | 1 | 649 | 94 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | thymocyte selection associated | Themis |
E430004N04Rik, Gasp, Spot, Tsepa, thylex |
-38.1kb | Anti-sense | Chr. 10 - 28668327 - 28883820 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 2310057J18 gene | 2310057J18Rik |
2210414P04Rik, Leg1 |
48.8kb | Sense | Chr. 10 - 28972288 - 28986306 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930519F09Rik
Genomic Map