This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04240883_s1
See other MIR1936 GE Assays ›
Gene Symbol:
Mir1936
Gene Name:
microRNA 1936
Gene Aliases:
mir-1936, mmu-mir-1936
Chromosome Location:
Chr.12: 102684928 - 102685020 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04240883_s1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer:
434.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mir1936
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_035457.1 1 39 63

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream inositol 1,3,4-triphosphate 5/6 kinase Itpk1 BC031182 Sense Chr. 12 - 102568583 - 102704869 on NCBI Build GRCm38
Downstream predicted gene 20604 Gm20604 AK010878-Moap1 54.8kb Sense Chr. 12 - 102739830 - 102757810 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200015662.mm.1 NR_035457.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200015662.mm.1 NR_035457.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200015662.mm.1 TC1200002254.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline