This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04278358_m1
See other GM9926 GE Assays ›
Gene Symbol:
Gm9926
Gene Name:
predicted gene 9926
Gene Aliases:
ENSMUSG00000053891
Chromosome Location:
Chr.18: 66504178 - 66511737 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04278358_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gm9926
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_040528.1 1 - 2 286 82
GenBank mRNA AK136461.1 - 1 - 2 287 82
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB624712.1 - 1 - 2 287 Chr. 18 - 66504178 - 66507515 [+] on Build GRCm38 82

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream phorbol-12-myristate-13-acetate-induced protein 1 Pmaip1 Noxa -38.6kb Sense Chr. 18 - 66458604 - 66465558 on NCBI Build GRCm38
Downstream melanocortin 4 receptor Mc4r Mc4-r 352.3kb Anti-sense Chr. 18 - 66857705 - 66860487 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1800000690.mm.1

Set Membership:

Amplicon length between 71 and 85 3' Most Mus musculus 1700 Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P106948 AK078927.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P173949 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P106948 AK078927.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P173949 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106948 AK078927.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P173949 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14031635 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13891675 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31787239 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31787236 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31787242 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31787241 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31787231 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14031630 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31787235 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13891678 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800005099.mm.1 NR_040528.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800005099.mm.1 NR_040528.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800005099.mm.1 TC1800000690.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1800005093.mm.1 NR_040528.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800005093.mm.1 NR_040528.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800005093.mm.1 TC1800000690.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1800005098.mm.1 NR_040528.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800005098.mm.1 NR_040528.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800005098.mm.1 TC1800000690.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1800005097.mm.1 NR_040528.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800005097.mm.1 NR_040528.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800005097.mm.1 TC1800000690.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1800005094.mm.1 NR_040528.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800005094.mm.1 NR_040528.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800005094.mm.1 TC1800000690.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1800005101.mm.1 NR_040528.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800005101.mm.1 NR_040528.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800005101.mm.1 TC1800000690.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1800005096.mm.1 NR_040528.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800005096.mm.1 NR_040528.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800005096.mm.1 TC1800000690.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1800005100.mm.1 NR_040528.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800005100.mm.1 NR_040528.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800005100.mm.1 TC1800000690.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 398145 - -
ABI Mouse Genome Survey Microarray 686468 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 104150184 - -
Illumina mouse-6 v1.0 expression beadchip 104150184 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1241279 - -
Illumina Sentrix mouse-6.v1 bead array 104150184 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 104150184 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1241279 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline