This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04335999_m1
See other MILR1 GE Assays ›
Gene Symbol:
Milr1
Gene Name:
mast cell immunoglobulin like receptor 1
Gene Aliases:
Allergin-1, Gm885, Mca32
Chromosome Location:
Chr.11: 106750652 - 106768794 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04335999_m1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer: 407.65
407.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Milr1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_073165.1 6 - 7 1118 93
GenBank mRNA AK137500.1 - 6 - 7 1077 93

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream polymerase (DNA directed),
gamma 2,
accessory subunit
Polg2 Anti-sense Chr. 11 - 106754821 - 106780010 on NCBI Build GRCm38
Downstream DEAD (Asp-Glu-Ala-Asp) box polypeptide 5 Ddx5 2600009A06Rik,
G17P1,
HUMP68,
Hlr1,
p68
16.4kb Anti-sense Chr. 11 - 106780355 - 106789268 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100001785.mm.1

Set Membership:

Amplicon length between 86 and 100 Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

immunoglobulin receptor superfamily

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of mast cell activation
mast cell degranulation
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100016862.mm.1 NM_001033435.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100016862.mm.1 NM_001271372.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100016862.mm.1 NM_001271373.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100016862.mm.1 NM_001271374.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100016862.mm.1 NM_001271375.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100016862.mm.1 NR_073165.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100016862.mm.1 TC1100001785.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0