This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04336353_g1
See other CAPG GE Assays ›
Gene Symbol:
Capg
Gene Name:
capping protein (actin filament), gelsolin-like
Gene Aliases:
gCap39, mbh1
Chromosome Location:
Chr.6: 72544391 - 72562983 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04336353_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Capg
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001042534.3 NP_001035999.1 6 - 7 913 115
NM_001271395.1 NP_001258324.1 6 - 7 906 115
NM_001271415.1 NP_001258344.1 7 - 8 903 115
NM_007599.4 NP_031625.2 6 - 7 760 115
GenBank mRNA AK146622.1 - 6 - 7 764 115
AK149857.1 - 6 - 7 710 115
AK152818.1 - 6 - 7 738 115
AK159577.1 - 6 - 7 713 115
AK159736.1 - 6 - 7 715 115
AK165153.1 - 6 - 7 727 115
AK165586.1 - 4 - 5 556 115
BC003480.1 - 6 - 7 714 2811050 115
BC023101.1 - 6 - 7 702 3983574 115
X54511.1 - 7 - 8 902 115

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Capg

Panther Classification:

Molecular Function -

non-motor actin binding protein

Gene Ontology Categories:

Function(s) Process(es)

cell projection assembly
barbed-end actin filament capping
actin filament capping
actin binding
protein domain specific binding
protein complex binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0