This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04337428_m1
See other ERCC8 GE Assays ›
Gene Symbol:
Ercc8
Gene Name:
excision repaiross-complementing rodent repair deficiency, complementation group 8
Gene Aliases:
2410022P04Rik, 2810431L23Rik, 4631412O06Rik, B130065P18Rik, Ckn1, Csa
Chromosome Location:
Chr.13: 108158694 - 108199284 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04337428_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ercc8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028042.3 NP_082318.2 4 - 5 454 113
GenBank mRNA AJ427222.1 - 4 - 5 398 113
AK010576.1 - 4 - 5 456 113
AK013216.1 - 4 - 5 454 112
AK014517.1 - 3 - 4 354 113
AK082109.1 - 3 - 4 344 113
AK082676.1 - 3 - 4 353 113
AK156267.1 - 4 - 5 442 113
AK157175.1 - 4 - 5 455 113
AK157504.1 - 3 - 4 353 113
BC037200.1 - 4 - 5 434 3985548 113
BC056463.1 - 3 - 4 336 113

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK157504.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ercc8

Panther Classification:

Molecular Function -

DNA metabolism protein nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
DNA repair
transcription-coupled nucleotide-excision repair
nucleotide-excision repair
cellular response to DNA damage stimulus
response to oxidative stress
response to UV
response to X-ray
proteasome-mediated ubiquitin-dependent protein catabolic process
positive regulation of DNA repair
protein autoubiquitination
ubiquitin-protein transferase activity
protein complex binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-48sk5:80/100.66.77.18:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0