This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04410411_m1
See other AI314278 GE Assays ›
Gene Symbol:
AI314278
Gene Name:
expressed sequence AI314278
Gene Aliases:
-
Chromosome Location:
Chr.7: 89426306 - 89432077 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04410411_m1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer: 360.22
360.22
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AI314278
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_102276.1 1 - 2 149 79
GenBank mRNA AK143758.1 - 1 - 2 147 79
AK165495.1 - 1 - 2 150 79
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY732584.1 - 1 - 2 147 Chr. 7 - 89426309 - 89430397 [+] on Build GRCm38 79
GenBankmRNA BG972953.1 - 1 - 2 118 Chr. 7 - 89426363 - 89430525 [+] on Build GRCm38 79
GenBankmRNA AW259824.1 - 1 - 2 86 Chr. 7 - 89426369 - 89430094 [+] on Build GRCm38 79
GenBankmRNA AI316901.1 - 1 - 2 141 Chr. 7 - 89426314 - 89430291 [+] on Build GRCm38 79
GenBankmRNA AW260645.1 - 1 - 2 86 Chr. 7 - 89426369 - 89429954 [+] on Build GRCm38 79
GenBankmRNA BY732867.1 - 1 - 2 150 Chr. 7 - 89426306 - 89430096 [+] on Build GRCm38 79

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream frizzled homolog 4 (Drosophila) Fzd4 Fz4 -13.2kb Sense Chr. 7 - 89404366 - 89413133 on NCBI Build GRCm38
Downstream protease,
serine 23
Prss23 2310046G15Rik,
4930479H08Rik,
AI790574,
Spuve
75.7kb Anti-sense Chr. 7 - 89507785 - 89527254 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-5xwv5:80/100.66.76.242:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0