This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04410706_m1
See other SNAP91 GE Assays ›
Gene Symbol:
Snap91
Gene Name:
synaptosomal-associated protein 91
Gene Aliases:
91kDa, AP180, F1-20, mKIAA0656
Chromosome Location:
Chr.9: 86765923 - 86880708 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04410706_m1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer: 360.22
360.22
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Snap91
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_102729.1 9 - 10 1140 72
GenBank mRNA BC052454.1 - 9 - 10 1140 6410678 72

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream protease,
serine 35
Prss35 6030424L22Rik,
P3D9
-63.3kb Anti-sense Chr. 9 - 86743240 - 86758443 on NCBI Build GRCm38
Downstream ripply2 homolog (zebrafish) Ripply2 C030002E08Rik,
Gm1122
190.6kb Anti-sense Chr. 9 - 87015128 - 87020441 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC052454.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

vesicle coat protein

Gene Ontology Categories:

Function(s) Process(es)

transport
establishment or maintenance of cell polarity
chemical synaptic transmission
neurotransmitter secretion
axonogenesis
protein transport
regulation of endocytosis
clathrin coat assembly
regulation of protein transport
regulation of clathrin-mediated endocytosis
protein binding
phospholipid binding
1-phosphatidylinositol binding
protein kinase binding
clathrin binding
clathrin heavy chain binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900023065.mm.1 NM_001277982.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900023065.mm.1 NM_001277983.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900023065.mm.1 NM_001277985.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900023065.mm.1 NM_001277986.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900023065.mm.1 NM_013669.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900023065.mm.1 NR_102729.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900023065.mm.1 TC0900002783.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-vvsgg:80/100.66.72.198:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0