Assay Details
| Gene Symbol: Slc10a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104094.1 | 1 - 2 | 348 | 104 | ||
| GenBank mRNA | AY825927.1 | - | 1 - 2 | 101 | 104 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
RNA binding motif protein, X linked-like-1 |
Rbmxl1 |
Hnrpg, Rbmx, Rbmxrt |
-400.0 | Anti-sense | Chr. 8 - 78505269 - 78508928 on NCBI Build GRCm38 |
| Downstream |
LSM6 homolog, U6 small nuclear RNA associated (S. cerevisiae) |
Lsm6 |
1500031N17Rik, 2410088K19Rik, AI747288 |
279.6kb | Anti-sense | Chr. 8 - 78804868 - 78821152 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Slc10a7
Genomic Map