This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04410801_m1
See other 1700029F12RIK GE Assays ›
Gene Symbol:
1700029F12Rik
Gene Name:
RIKEN cDNA 1700029F12 gene
Gene Aliases:
-
Chromosome Location:
Chr.13: 97021864 - 97035254 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04410801_m1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer: 407.65
407.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 1700029F12Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_104276.1 1 - 2 35 73
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CB274252.1 - 1 - 2 35 Chr. 13 - 97030275 - 97035254 [-] on Build GRCm38 73

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream glucosaminyl (N-acetyl) transferase 4,
core 2 (beta-1,6-N-acetylglucosaminyltransferase)
Gcnt4 C2gnt3,
Gm279,
Gm73
-79.3kb Anti-sense Chr. 13 - 96924352 - 96950914 on NCBI Build GRCm38
Downstream family with sequence similarity 169,
member A
Fam169a B230112C05Rik,
SLAP75
32.0kb Anti-sense Chr. 13 - 97067287 - 97129977 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300002475.mm.1

Set Membership:

Amplicon length between 71 and 85 Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300016620.mm.1 NM_001080777.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300016620.mm.1 NM_001284205.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300016620.mm.1 NM_025585.3 Refseq
Affymetrix Mouse Clariom S Array PSR1300016620.mm.1 NR_104276.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300016620.mm.1 TC1300002475.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300016624.mm.1 NR_104276.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300016624.mm.1 TC1300002475.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0