This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04411929_g1
See other STAU1 GE Assays ›
Gene Symbol:
Stau1
Gene Name:
staufen (RNA binding protein) homolog 1 (Drosophila)
Gene Aliases:
5830401L18Rik, AW549911, C85792, Stau
Chromosome Location:
Chr.2: 166947549 - 166996299 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04411929_g1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Stau1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_111895.1 8 - 9 1162 76
GenBank mRNA AY391773.1 - 8 - 9 1161 76

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream chromosome segregation 1-like (S. cerevisiae) Cse1l 2610100P18Rik,
AA407533,
Capts,
Cas,
Xpo2
-4.4kb Anti-sense Chr. 2 - 166906051 - 166946389 on NCBI Build GRCm38
Downstream DEAD (Asp-Glu-Ala-Asp) box polypeptide 27 Ddx27 C86129 64.2kb Anti-sense Chr. 2 - 167015255 - 167034950 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

intracellular mRNA localization
positive regulation of viral genome replication
positive regulation by virus of viral protein levels in host cell
RNA binding
double-stranded RNA binding
protein binding
protein phosphatase 1 binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-x8dcn:80/100.66.79.134:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0