This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04418570_g1
See other OAZ2 GE Assays ›
Gene Symbol:
Oaz2
Gene Name:
ornithine decarboxylase antizyme 2
Gene Aliases:
AZ-2, AZ2, Oaz2-ps, Sez15
Chromosome Location:
Chr.9: 65676548 - 65690300 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04418570_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Oaz2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001301307.1 NP_001288236.1 1 - 2 220 95
GenBank mRNA CJ072367.1 - 1 - 2 190 95

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001301307.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

protein-binding activity modulator

Gene Ontology Categories:

Function(s) Process(es)

polyamine metabolic process
polyamine biosynthetic process
negative regulation of catalytic activity
positive regulation of protein catabolic process
positive regulation of intracellular protein transport
negative regulation of polyamine transmembrane transport
enzyme inhibitor activity
protein binding
ornithine decarboxylase inhibitor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0