This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04419769_s1
See other MGMT GE Assays ›
Gene Symbol:
Mgmt
Gene Name:
O-6-methylguanine-DNA methyltransferase
Gene Aliases:
AGT, AI267024, Agat
Chromosome Location:
Chr.7: 136894468 - 137128193 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04419769_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mgmt
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008598.2 NP_032624.1 5 770 104
GenBank mRNA AK090389.1 - 760 104
BC096557.1 - 5 773 1366095 104
M84524.1 - 4 696 104

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mgmt

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
DNA dealkylation involved in DNA repair
cellular response to DNA damage stimulus
methylation
regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of DNA repair
negative regulation of cell death
DNA binding
damaged DNA binding
catalytic activity
methylated-DNA-[protein]-cysteine S-methyltransferase activity
calcium ion binding
methyltransferase activity
transferase activity
metal ion binding

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 860325 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0