This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04461891_m1
See other ATF2 GE Assays ›
Gene Symbol:
Atf2
Gene Name:
activating transcription factor 2
Gene Aliases:
Atf-2, CRE-BP, Creb2, D130078H02Rik, D18875, Tg(Gzma-Klra1)7Wum, mXBP
Chromosome Location:
Chr.2: 73816509 - 73892639 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04461891_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atf2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001025093.2 NP_001020264.1 10 - 11 1108 96
NM_001284369.1 NP_001271298.1 8 - 9 948 96
NM_001284370.1 NP_001271299.1 10 - 11 1108 96
NM_001284371.1 NP_001271300.1 9 - 10 1045 96
NM_009715.3 NP_033845.1 9 - 10 1011 96
GenBank mRNA AF483482.1 - 6 - 7 685 96
AF483483.1 - 6 - 7 685 96
AK035438.1 - 10 - 11 1064 96
AK041145.1 - 4 - 5 2688 96
AK051591.1 - 9 - 10 1001 96
AK139515.1 - 8 - 9 864 96
AK139844.1 - 8 - 9 861 96
AK146839.1 - 9 - 10 957 96
AY902311.1 - 5 - 6 571 96
BC042210.1 - 7 - 8 784 4018323 96
M77167.1 - 4 - 5 457 96
S76657.1 - 776 96

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Atf2

Panther Classification:

Molecular Function -

basic leucine zipper transcription factor

Gene Ontology Categories:

Function(s) Process(es)

outflow tract morphogenesis
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
response to osmotic stress
cellular response to DNA damage stimulus
histone acetylation
intra-S DNA damage checkpoint
positive regulation of transforming growth factor beta2 production
fat cell differentiation
positive regulation of transcription from RNA polymerase II promoter
positive regulation of sequence-specific DNA binding transcription factor activity
adipose tissue development
positive regulation of mitochondrial membrane permeability involved in apoptotic process
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II activating transcription factor binding
enhancer sequence-specific DNA binding
nucleic acid binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
histone acetyltransferase activity
protein binding
cAMP response element binding protein binding
transferase activity
protein kinase binding
cAMP response element binding
sequence-specific DNA binding
metal ion binding
protein heterodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline