This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04933133_m1
See other PEMT GE Assays ›
Gene Symbol:
Pemt
Gene Name:
phosphatidylethanolamine N-methyltransferase
Gene Aliases:
PEAMT, PEMT2, Pempt, Pempt2
Chromosome Location:
Chr.11: 59970614 - 60046512 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04933133_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pemt
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001290014.1 NP_001276943.1 3 - 4 305 74

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

methyltransferase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
phospholipid metabolic process
glycerophospholipid metabolic process
phosphatidylcholine biosynthetic process
negative regulation of cell proliferation
phospholipid biosynthetic process
methylation
response to vitamin
response to drug
S-adenosylhomocysteine metabolic process
S-adenosylmethionine metabolic process
positive regulation of lipoprotein metabolic process
positive regulation of protein targeting to mitochondrion
phosphatidyl-N-methylethanolamine N-methyltransferase activity
phosphatidylethanolamine N-methyltransferase activity
methyltransferase activity
N-methyltransferase activity
phosphatidylethanolamine binding
S-adenosylmethionine-dependent methyltransferase activity
transferase activity
phosphatidyl-N-dimethylethanolamine N-methyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline