This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04933208_m1
See other ATP5J GE Assays ›
Gene Symbol:
Atp5j
Gene Name:
ATP synthase, H+ transporting, mitochondrial F0 complex, subunit F
Gene Aliases:
CF6
Chromosome Location:
Chr.16: 84827866 - 84835648 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04933208_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atp5j
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AA259963.1 - 3 - 4 336 122
AK078484.1 - 3 - 4 497 122
BC010766.1 - 4 - 5 632 4219715 122
BE851144.1 - 4 - 5 374 122
U77128.1 - 2 - 3 295 122
W75852.1 - 3 - 4 362 122

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

ATP synthase

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30925871 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30925872 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30925875 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30925869 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1963772 NR_002690.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 793110 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 338171 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 470279 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 720090 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 57854 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P154471 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1963772 NR_002690.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P100866 AK078484.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P100866 BC010766.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P100866 U77128.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P100866 NM_016755.2 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30925891 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30925884 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30925872 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30925879 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30925871 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30925897 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30925875 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30925886 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30925895 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30925896 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30925890 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30925873 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30925881 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30925878 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30925892 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30925883 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30925899 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30925870 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30925887 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08667315 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P154471 - -
Whole Mouse Genome G4122A A_51_P100866 AK078484.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P100866 BC010766.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P100866 U77128.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P100866 NM_016755.2 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119351 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115224 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P154471 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12923791 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12524196 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30925869 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30925871 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30925875 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30925872 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30925911 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30925892 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30925913 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30925875 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30925908 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30925883 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30925887 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30925919 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30925905 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30925822 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30925871 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30925900 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30925918 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30925895 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30925806 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30925820 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30925817 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30925878 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30925886 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30925903 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30925926 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30925927 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30925897 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30925910 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30925805 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30925881 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30925890 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30925811 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30925873 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30925899 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30925906 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30925923 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30925896 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30925884 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30925814 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30925921 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30925915 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30925808 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30925907 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30925929 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30925821 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30925821 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30925877 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30925896 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30925808 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30925900 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12943097 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30925905 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30925809 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30925805 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12538183 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30925895 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30925886 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30925887 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30925910 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12959087 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30925883 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30925899 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30925890 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30925911 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30925913 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30925820 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30925891 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13022696 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30925806 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30925908 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30925893 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30925903 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30925892 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30925906 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30925884 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12521612 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30925880 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30925875 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10440487 - -
Affymetrix Mouse Gene 1.0 ST Array 10440485 - -
Affymetrix Mouse Gene 1.0 ST Array 10440484 - -
Affymetrix Mouse Gene 1.0 ST Array 10440486 - -
Affymetrix HT MG-430 PM Array Plate 1416143_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10440483 - -
Mouse Genome 430 2.0 Array 1416143_PM_at - -
Mouse Genome 430 2.0 Array 1416143_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 11957_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 11957_PM_at - -
Affymetrix Mouse Clariom S Array PSR1600015219.mm.1 NM_016755.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600015219.mm.1 TC1600001964.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600015197.mm.1 NM_016755.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600015197.mm.1 TC1600001964.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600015221.mm.1 NM_016755.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600015221.mm.1 TC1600001964.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600015212.mm.1 NM_016755.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600015212.mm.1 TC1600001964.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600015199.mm.1 NM_016755.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600015199.mm.1 TC1600001964.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600015213.mm.1 NM_016755.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600015213.mm.1 TC1600001964.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600015203.mm.1 NM_016755.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600015203.mm.1 TC1600001964.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600015214.mm.1 NM_016755.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600015214.mm.1 TC1600001964.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600015204.mm.1 NM_016755.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600015204.mm.1 TC1600001964.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600015220.mm.1 NM_016755.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600015220.mm.1 TC1600001964.mm.1 MTA TC
Mouse Expression Set 430 1416143_at - -
Mouse Expression Set 430 1416143_PM_at - -
Murine Genome U74v2 Set 98128_PM_at - -
Murine Genome U74v2 Set 98128_at - -
Mouse Genome 430A 2.0 Array 1416143_at - -
Mouse Genome 430A 2.0 Array 1416143_PM_at - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 4210100 - -
Illumina mouse-6 v1.0 expression beadchip 6900288 - -
Illumina mouseRef-8 v1.0 expression beadchip 0004210100 - -
Illumina mouseRef-8 v1.0 expression beadchip 0006900288 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1231207 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2632890 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1231207 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1212748 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2632890 - -
mouse-6 v1.1 (Illumina) scl0000102.1_16-S - -
mouse-6 v1.1 (Illumina) scl48278.6.1_15-S - -
Illumina Sentrix mouse-6.v1 bead array 4210100 - -
Illumina Sentrix mouse-6.v1 bead array 6900288 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl48278.6.1_15-S - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl0000102.1_16-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 4210100 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 6900288 - -
MouseRef-8 v2 Expression BeadChip ILMN_2611261 - -
MouseRef-8 v2 Expression BeadChip ILMN_2632890 - -
MouseRef-8 v2 Expression BeadChip ILMN_2962632 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2962632 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2632890 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2611261 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0000102.1_16-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0