Assay Details
| Gene Symbol: Rny3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024202.2 | 1 | 52 | 71 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | 4.5S RNA | Rn4.5s | -126.5kb | Anti-sense | Chr. 6 - 47654921 - 47655094 on NCBI Build GRCm38 | |
| Downstream |
RNA, Y1 small cytoplasmic, Ro-associated |
Rny1 | 6.3kb | Anti-sense | Chr. 6 - 47788070 - 47788180 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Rny3
Genomic Map