This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04934564_m1
See other ETFB GE Assays ›
Gene Symbol:
Etfb
Gene Name:
electron transferring flavoprotein, beta polypeptide
Gene Aliases:
0610009I16Rik, 2810441H06Rik
Chromosome Location:
Chr.7: 43444072 - 43457800 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04934564_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Etfb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026695.3 NP_080971.2 1 - 2 129 95
NR_075104.1 1 - 2 129 95
GenBank mRNA AK002407.1 - 1 - 2 71 95
AK013284.1 - 1 - 2 94 95
AK150293.1 - 1 - 2 72 95
BC049237.1 - 1 - 2 58 4241552 95
BC069877.1 - 1 - 2 88 5370580 95
BY292045.1 - 1 - 2 130 95

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Etfb

Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

transport
oxidation-reduction process
electron carrier activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-6s775:80/100.66.79.102:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0