This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04934565_m1
See other ETFB GE Assays ›
Gene Symbol:
Etfb
Gene Name:
electron transferring flavoprotein, beta polypeptide
Gene Aliases:
0610009I16Rik, 2810441H06Rik
Chromosome Location:
Chr.7: 43444072 - 43457800 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04934565_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Etfb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026695.3 NP_080971.2 2 - 3 294 100
NR_075104.1 2 - 3 294 100
GenBank mRNA AA207939.1 - 2 - 3 178 100
AK002407.1 - 2 - 3 236 100
AK150293.1 - 2 - 3 237 100
BC049237.1 - 2 - 3 223 4241552 100
BC069877.1 - 2 - 3 253 5370580 100
BY292045.1 - 2 - 3 296 101

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Etfb

Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

transport
oxidation-reduction process
electron carrier activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hk65g:80/100.66.77.69:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0