This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm05700208_s1
See other TRAPPC2 GE Assays ›
Gene Symbol:
Trappc2
Gene Name:
trafficking protein particle complex 2
Gene Aliases:
1110066L09Rik, 1810064C02Rik, AW496358, MIP-2A, Sedl, TRS20
Chromosome Location:
Chr.X: 166440825 - 166452543 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm05700208_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Trappc2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025432.3 NP_079708.2 2 156 87
GenBank mRNA AK002658.1 - 3 227 87
AK004389.1 - 181 87
AK007889.1 - 2 157 87
AK007951.1 - 206 87
AK008564.1 - 3 206 87
AK034967.1 - 3 207 87
AK077643.1 - 182 87
AY318751.1 - 214 87
BC034845.1 - 191 1528500 87
BC061087.1 - 226 6589013 87
BC099551.1 - 198 30252145 87

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Amplicon spans exons and probe does not span exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Trappc2

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

skeletal system development
regulation of transcription, DNA-templated
protein binding
transcription factor binding
ion channel binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-bjcbn:80/100.66.75.54:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0